Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triolein ≥75%, tech.

Supplier: Thermo Scientific Chemicals
Synonyms: Glycerol Trioleate

J62419-22 J62419-22 J62419-14 J62419-14
AAJ62419-14EA 162.32 USD InStock
AAJ62419-14 AAJ62419-22 CAAAJ62419-14 CAAAJ62419-22
Triolein ≥75%, tech.
Triolein
Liquid


Formula: C₅₇H₁₀₄O₆
MW: 885.45 g/mol
Boiling Pt: 235…240 °C/18 mm
Density: 0,91
Flash Pt: 330 °C(626 °F)
Storage Temperature: Freezer
MDL Number: MFCD00137563
CAS Number: 122-32-7
EINECS: 204-534-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty