Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Penicillin V potassium salt

Supplier: Thermo Scientific Chemicals
Danger

J62442-14 J62442-22 J62442-14 J62442-09 J62442-09 J62442-22
AAJ62442-14EA 93.44 USD InStock
CAAAJ62442-09 AAJ62442-14 AAJ62442-22 CAAAJ62442-14 AAJ62442-09 CAAAJ62442-22
Penicillin V potassium salt
Penicillin V potassium salt
Inhibits bacterial cell wall synthesis

White to off-white powder


Formula: C₁₆H₁₇KN₂O₅S
MW: 388.48 g/mol
Melting Pt: 197…202 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051771
CAS Number: 132-98-9
EINECS: 205-086-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty