Synonyms:
9,10-deepithio-9,10-didehydroacanthifolicin sodium salt
, OA sodium salt
J62463-EXS
AAJ62463-EXSEA
587.43
USD
InStock
AAJ62463-EXS
Okadaic acid sodium salt 98%, Powder
Okadaic acid sodium salt
Potent inhibitor of serine/threonine-specific protein phosphatases 1 and 2A (PP1 and PP2A). Does not inhibit tyrosine or alkaline phosphatases