Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Paraformaldehyde 4% in PBS + Mg + EGTA

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Polyoxymethylene

J62478-AP J62478-AK J62478-AK J62478-AP
AAJ62478-AKEA 173.03 USD InStock
CAAAJ62478-AK CAAAJ62478-AP AAJ62478-AK AAJ62478-AP
Paraformaldehyde 4% in PBS + Mg + EGTA
Paraformaldehyde
For histology.

Liquid


Formula: (CH₂O)n
Storage Temperature: Refrigerator
MDL Number: MFCD00133991
CAS Number: 30525-89-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty