Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Galantamine hydrobromide

Supplier: Thermo Scientific Chemicals
Danger

J62482-MF J62482-03 J62482-MF J62482-03
AAJ62482-MFEA 123.77 USD InStock
AAJ62482-MF CAAAJ62482-MF AAJ62482-03 CAAAJ62482-03
Galantamine hydrobromide
Galantamine hydrobromide
Centrally active and long-acting acetylcholinesterase inhibitor

Powder

Soluble in water or DMSO


Formula: C₁₇H₂₁NO₃·HBr
MW: 368.27 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00067672
CAS Number: 1953-04-4
EINECS: 217-780-5
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty