Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TRIS-buffered saline (TBS, 10X), with 1% Triton® X-100, Thermo Scientific

Supplier: Thermo Scientific
J62533-K2 J62533-AP J62533-AP J62533-AK
AAJ62533-APEA 73.24 USD InStock
AAJ62533-AP CAAAJ62533-K2 AAJ62533-AK CAAAJ62533-AP
TRIS-buffered saline (TBS, 10X), with 1% Triton® X-100, Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers
Liquid

Physical Form: Liquid
MDL No.: MFCD00132476

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty