Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form) 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-((2R,3R,4S,5R)-5-((((((((2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium, disodium saltNADP disodium saltTriphosphopyridine nucleotide, disodium saltNicotinamide-adenine dinucleotide phosphate disodium saltNADP-Na2β-NADP-Na2Nadide phosphate disodium saltβ-NADH phosphate disodium salt

J62556-03 J62556-MD J62556-MD J62556-03
AAJ62556-03EA 387.4 USD InStock
CAAAJ62556-03 CAAAJ62556-MD AAJ62556-03 AAJ62556-MD
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form) 97%
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Powder


Formula: C₂₁H₂₆N₇Na₂O₁₇P₃
MW: 787.37 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065390
CAS Number: 24292-60-2
EINECS: 246-129-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty