Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(+)-1-Deoxynojirimycin

Supplier: Thermo Scientific Chemicals

J62602-MC J62602-MC
AAJ62602-MCEA 593.79 USD InStock
AAJ62602-MC CAAAJ62602-MC
(+)-1-Deoxynojirimycin
(+)-1-Deoxynojirimycin
An alpha-glucosidase inhibitor. Interferes with normal processing of N-linked glycoproteins.

Powder


Formula: C₆H₁₃NO₄
MW: 163.17
Melting Pt: 195…196 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063474
CAS Number: 19130-96-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty