Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-thyroxine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid

J62606-06 J62606-03 J62606-06 J62606-03
AAJ62606-03EA 82.15 USD InStock
CAAAJ62606-06 CAAAJ62606-03 AAJ62606-03 AAJ62606-06
L-thyroxine 98%
L-thyroxine
Crystalline powder


Formula: C₁₅H₁₁I₄NO₄
MW: 776.874 g/mol
Melting Pt: 235…236 °C
Storage Temperature: Ambient
MDL Number: MFCD00002595
CAS Number: 51-48-9
EINECS: 200-101-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty