Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sorbitan sesquioleate

Supplier: Thermo Scientific Chemicals
Synonyms: Arlacel 83Arlacel CSorbitan Sesquioleate

J62648-AP J62648-K2 J62648-K2 J62648-AP
AAJ62648-K2EA 96.57 USD InStock
AAJ62648-K2 CAAAJ62648-AP AAJ62648-AP CAAAJ62648-K2
Sorbitan sesquioleate
Sorbitan sesquioleate
An emulsifier

Viscous liquid


Formula: C₄₂H₇₆O₇
MW: 693.06 g/mol
Density: 0.989
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00151568
CAS Number: 8007-43-0
EINECS: 232-360-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty