Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

G418 disulfate cell culture reagent

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: G418 disulfate G-418 disulfate Geneticin® G-418 sulfate

J62671-14 J62671-03 J62671-06 J62671-03 J62671-06
AAJ62671-06EA 572.61 USD InStock
AAJ62671-06 CAAAJ62671-06 CAAAJ62671-03 AAJ62671-14 AAJ62671-03
G418 disulfate cell culture reagent
G418 disulphate
Aminoglycoside that is toxic to bacteria, yeast, protozoans and helminths

Powder


Formula: C₂₀H₄₄N₄O₁₈S₂
MW: 692.72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058314
CAS Number: 108321-42-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty