Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TE buffer, pH 8.1, RNAse free, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

J62745-EQE J62745-AK J62745-AK J62745-EQE
AAJ62745-AKEA 197.22 USD InStock
AAJ62745-AK CAAAJ62745-EQE AAJ62745-EQE CAAAJ62745-AK
TE buffer, pH 8.1, RNAse free, Thermo Scientific
Buffers Cell Culture Buffers
Liquid

Physical Form: Liquid
MDL No.: MFCD00236359

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty