Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Asparagine monohydrate Cell Culture Reagent

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-(+)-Asparagine monohydrate H-Asn-OH.H2O

J62869-22 J62869-A1 J62869-36 J62869-22 J62869-A1 J62869-36
AAJ62869-22EA 95.56 USD InStock
AAJ62869-22 AAJ62869-A1 AAJ62869-36 CAAAJ62869-36 CAAAJ62869-22 CAAAJ62869-A1
L(+)-Asparagine monohydrate Cell Culture Reagent
L(+)-Asparagine monohydrate
White to off-white


Formula: C₄H₈N₂O₃·1H₂O
MW: 150.13 g/mol
Melting Pt: 215…217 °C
Storage Temperature: Ambient
MDL Number: MFCD00151038
CAS Number: 5794-13-8
EINECS: 200-735-9
Merck Index: 13,00842
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty