Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Methionine sulfoxide

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (2S)-2-Amino-4-(methylsulfinyl)butanoic acidH-Met(O)-OH(2S)-2-Amino-4-(methylsulphinyl)butanoic acid

J62873-03 J62873-14 J62873-06 J62873-14 J62873-03 J62873-06
AAJ62873-06EA 182.34 USD InStock
AAJ62873-06 AAJ62873-14 AAJ62873-03 CAAAJ62873-06 CAAAJ62873-03 CAAAJ62873-14
L-Methionine sulfoxide
L-Methionine sulphoxide
Powder


Formula: C₅H₁₁NO₃S
MW: 165.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063093
CAS Number: 3226-65-1
EINECS: 221-758-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty