Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

U-73122 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-[6-[((17beta)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]pyrrole-2,5-dione

J62898-MA J62898-MA J62898-MCR J62898-LB0 J62898-LB0 J62898-MCR
AAJ62898-MAEA 257.3 USD InStock
CAAAJ62898-LB0 AAJ62898-MA AAJ62898-LB0 CAAAJ62898-MCR CAAAJ62898-MA AAJ62898-MCR
U-73122 95%
U-73122
Inhibitor of phospholipase C activation and inhibits the down regulation of muscarinic receptors

Crystalline solid

Soluble in DMSO, ethanol, or DMF at approx. 0.5mg/ml. May require heating or overnight mixing. Insoluble in water


Formula: C₂₉H₄₀N₂O₃
MW: 464.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00893825
CAS Number: 112648-68-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty