Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA (ethylenediamine tetraacetic acid) cell culture reagent

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ethylenediaminetetraacetic acid2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acidIdranal® IIKestranal® AEdetic AcidEDTAEdetic acidEthylenedinitrilotetra-acetic acid

J62948-36 J62948-36 J62948-A1 J62948-A1
AAJ62948-A1EA 122.72 USD InStock
AAJ62948-A1 AAJ62948-36 CAAAJ62948-36 CAAAJ62948-A1
EDTA (ethylenediamine tetraacetic acid) cell culture reagent
EDTA (Ethylenediamine tetraacetic acid)
Powder


Formula: C₁₀H₁₆N₂O₈
MW: 292.25 g/mol
Melting Pt: ca 245 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/41/2297241.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty