Would you like to log in again?
Bovine Superoxide dismutase (from Erythrocytes)
Supplier: Thermo Scientific
J63003-MA J63003-8PL J63003-8PL J63003-MA
AAJ63003-MAEA 362.76 USD InStock
CAAAJ63003-MA AAJ63003-MA AAJ63003-8PL CAAAJ63003-8PL
Bovine Superoxide dismutase (from Erythrocytes)
Enzymes
Catalyzes the dismutation of superoxide radicals to hydrogen peroxide and molecular oxygen.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty