Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Naphthyl phosphate monosodium salt monohydrate 97+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Monosodium 1-Naphthyl Phosphate MonohydrateSodium 1-Naphthyl Hydrogen Phosphate Monohydrate1-Naphthylphosphoric Acid Monosodium Salt MonohydratePhosphoric Acid 1-Naphthyl Ester Monosodium Salt Monohydrate

J63004-14 J63004-14 J63004-06 J63004-06
AAJ63004-14EA 411.85 USD InStock
CAAAJ63004-14 AAJ63004-14 AAJ63004-06 CAAAJ63004-06
1-Naphthyl phosphate monosodium salt monohydrate 97+%
1-Naphthyl phosphate monosodium salt monohydrate
Substrate for determination of phosphatases.

Powder


Formula: C₁₀H₈NaO₄P·1H₂O
MW: 264.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150615
CAS Number: 81012-89-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty