Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

DNA Marker, High Range, 1000 bp, Thermo Scientific

Supplier: Thermo Scientific
J63178-S0 J63178-XF J63178-S0 J63178-XF
AAJ63178-S0EA 212.03 USD InStock
CAAAJ63178-S0 CAAAJ63178-XF AAJ63178-S0 AAJ63178-XF
DNA Marker, High Range, 1000 bp, Thermo Scientific
Electrophoresis Reagents Molecular Weight Markers
DNA ladders are used in gel electrophoresis to determine the size and quantity of testing DNA fragments of genomic, plasmid, and PCR DNA.

  • Form: Liquid
  • Storage: Store at –20 °C
  • Solubility: Soluble in 10 mM Tris-HCl (pH 8.0), 1.0 mM EDTA, 50 mM NaCl
  • Contains 8 bands from 1000 to 10000 base pairs: 1000 bp increments from 1000 to 6000 bp, 8000 and 10000 bp

It is commonly used in the studies of plasmid DNA and PCR products. 1 kb DNA Ladder has been used as a molecular marker to determine the molecular weight and size of double stranded DNA during electrophoresis. 1 kb Ladder contains 11 fragments consisting of 500 bp repeats from 0.5 to 3 kb, 1 kb repeats from 3 to 6 kb, and 2 kb repeats from 6 to 10 kb. In NA electrophoresis, five of the marker should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel. Suitable for use in northern and southern blotting. All DNA markers can be stained with ethidium bromide, SYBR® Green, and Nancy-520.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty