Would you like to log in again?
YT Microbial Medium, 2X, Thermo Scientific
Supplier: Thermo Scientific
J63227-K2 J63227-AP J63227-AP J63227-K2
AAJ63227-APEA 94.03 USD InStock
CAAAJ63227-AP AAJ63227-AP CAAAJ63227-K2 AAJ63227-K2
YT Microbial Medium, 2X, Thermo Scientific
Cell Culture Media
Growth medium for culturing Escherichia coli, particularly laboratory or recombinant strains.
  • Liquid
Ingredients (per liter amounts): 16 g tryptone (pancreatic digest of casein), 10 g yeast extract, 5 g NaCl
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty