Would you like to log in again?
Clostridium perfringens Neuraminidase
Supplier: Thermo Scientific
J63259-MA J63259-MA
AAJ63259-MAEA 401.83 USD InStock
AAJ63259-MA CAAAJ63259-MA
Clostridium perfringens Neuraminidase
Enzymes
Applications: A hydrolytic enzyme that removes sialic acid from mucoproteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty