Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cytidine-5'-monophosphate disodium salt ≥99%

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium ((2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate

J63376-14 J63376-14 J63376-06 J63376-06 J63376-03 J63376-03
AAJ63376-14EA 363.1 USD InStock
CAAAJ63376-06 AAJ63376-14 AAJ63376-03 AAJ63376-06 CAAAJ63376-14 CAAAJ63376-03
Cytidine-5'-monophosphate disodium salt ≥99%
Cytidine-5'-monophosphate disodium salt
Crystalline powder


Formula: C₉H₁₂N₃Na₂O₈P
MW: 367.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064355
CAS Number: 6757-06-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty