Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zeatin riboside

Supplier: Thermo Scientific Chemicals
Synonyms: trans-Zeatinribosidetrans-Zeatin-riboside

J63382-MF J63382-MF J63382-MB J63382-MB J63382-MC J63382-MC
AAJ63382-MFEA 333.69 USD InStock
AAJ63382-MF CAAAJ63382-MF CAAAJ63382-MC CAAAJ63382-MB AAJ63382-MB AAJ63382-MC
Zeatin riboside
Zeatin riboside
Main effects in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence

Powder


Formula: C₁₅H₂₁N₅O₅
MW: 351.363 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036809
CAS Number: 6025-53-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty