Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gellan gum

Supplier: Thermo Scientific Chemicals

J63423-30 J6342330 J63423-A1 J63423-A1
AAJ63423-30EA 164.28 USD InStock
CAAAJ63423-A1 CAAAJ63423-30 AAJ63423-30 AAJ63423-A1
Gellan gum
Gellan gum
An agar substitute and gelling agent

Powder


Storage Temperature: Ambient
EINECS: 275-117-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty