Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Substance P

Supplier: Thermo Scientific
J63489-MB J63489-LB0 J63489-LB0 J63489-MB
AAJ63489-LB0EA 180.58 USD InStock
AAJ63489-LB0 CAAAJ63489-LB0 CAAAJ63489-MB AAJ63489-MB
Substance P
Proteins and Peptides
Powder
A neurokinin NK-1 receptor agonist

CAS No.: 33507-63-0
Molecular Formula: C63H98N18O13S
Formula Weight: 1347.64
Physical Form: Powder
Appearance: White

A neurokinin NK-1 receptor agonist

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty