Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Imidazole, 0.5 M Buffer Solution, pH 7.5, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

J63496-AE J63496-AE J63496-AK J63496-AK
AAJ63496-AEEA 153.6 USD InStock
CAAAJ63496-AE AAJ63496-AE CAAAJ63496-AK AAJ63496-AK
Imidazole, 0.5 M Buffer Solution, pH 7.5, Thermo Scientific
Buffers Biological Buffers
Liquid

CAS No.: 288-32-4
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
MDL No.: MFCD00005183

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty