Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bombesin

Supplier: Thermo Scientific
J63514-MCR J63514-LB0 J63514-MCR J63514-LB0
AAJ63514-LB0EA 208.75 USD InStock
CAAAJ63514-LB0 CAAAJ63514-MCR AAJ63514-LB0 AAJ63514-MCR
Bombesin
Proteins and Peptides
CAS No.: 31362-50-2
Molecular Formula: C71H110N24O18S
Formula Weight: 1619.90
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in 0.05M acetic acid (20mg/ml)
Merck Reference: 14,1329
MDL No.: MFCD00167514

Gut tetradecapeptide with the ability to stimulate release of various hormones

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty