Would you like to log in again?
Prehybridization Solutions in SSC, Thermo Scientific
Supplier: Thermo Scientific
J63618-AD J63618-AE J63618-AE J63618-AD
AAJ63618-ADEA 63.1 USD InStock
AAJ63618-AD AAJ63618-AE CAAAJ63618-AD CAAAJ63618-AE
Prehybridization Solutions in SSC, Thermo Scientific
Buffers Electrophoresis Buffers Hybridization Buffers
For the hybridization of probes to nucleic acids in Nothern and Southern blotting.
  • Liquid
Delivery information: Contains 6X SSC, 5X Denhardt's solution, 0.5% SDS, and 100 ug/ml of denatured DNA from salmon testes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty