Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Thio-D-glucose ≥97%

Supplier: Thermo Scientific Chemicals

J63621-MB J63621-LB0 J63621-MB
AAJ63621-MBEA 189.71 USD InStock
CAAAJ63621-LB0 CAAAJ63621-MB AAJ63621-MB
5-Thio-D-glucose ≥97%
5-Thio-D-glucose
Powder


Formula: C₆H₁₂O₅S
MW: 196.22 g/mol
Melting Pt: 135…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00006663
CAS Number: 20408-97-3
EINECS: 243-798-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty