Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(hydroxymethyl)aminomethane Hydrochloride, 1M stock soln, pH 8.0, Thermo Scientific

Supplier: Thermo Scientific
J63636-K2 J63636-K2
AAJ63636-K2EA 165.94 USD InStock
AAJ63636-K2 CAAAJ63636-K2
Tris(hydroxymethyl)aminomethane Hydrochloride, 1M stock soln, pH 8.0, Thermo Scientific
Buffers Cloning Buffers
Liquid

CAS No.: 1185-53-1
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00012590

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty