Would you like to log in again?
Citrate-Buffered Saline (20X), Thermo Scientific
Supplier: Thermo Scientific
J63666-K2 J63666-K3 J63666-K3 J63666-K2
AAJ63666-K2EA 40.77 USD InStock
AAJ63666-K2 CAAAJ63666-K3 AAJ63666-K3 CAAAJ63666-K2
Citrate-Buffered Saline (20X), Thermo Scientific
Buffers Electrophoresis Buffers Hybridization Buffers
Saline-sodium citrate (SSC) buffer.
300 mM sodium citrate, 3M sodium chloride, pH 7.0
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty