Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Threonine Cell Culture Reagent

Supplier: Thermo Scientific Chemicals
Synonyms: (2S,3R)-(-)-Threonine(2S,3R)-2-Amino-3-hydroxybutyric acidH-Thr-OH

J63709-30 J63709-A1 J63709-A1 J63709-30
AAJ63709-A1EA 209.95 USD InStock
AAJ63709-A1 AAJ63709-30 CAAAJ63709-30 CAAAJ63709-A1
L-Threonine Cell Culture Reagent
L-Threonine
Powder


Formula: C₄H₉NO₃
MW: 119.12 g/mol
Melting Pt: 256 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064270
CAS Number: 72-19-5
EINECS: 200-774-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty