Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Cysteine Cell Culture Reagent

Supplier: Thermo Scientific Chemicals
Synonyms: H-Cys-OH(R)-(+)-Cysteine(R)-2-Amino-3-mercaptopropanoic acid

J63745-22 J63745-14 J63745-14 J63745-22
AAJ63745-22EA 125.83 USD InStock
AAJ63745-22 AAJ63745-14 CAAAJ63745-14 CAAAJ63745-22
L(+)-Cysteine Cell Culture Reagent
L(+)-Cysteine
Powder


Formula: HSCH₂CH(NH₂)CO₂H
MW: 121.16 g/mol
Melting Pt: ca 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064306
CAS Number: 52-90-4
EINECS: 200-158-2
Merck Index: 13,02810
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty