Would you like to log in again?
Yeast Hexokinase
Supplier: Thermo Scientific
J63795-JPL J63795-4I J63795-JPL J63795-4I
AAJ63795-4IEA 147.65 USD InStock
CAAAJ63795-4I AAJ63795-4I AAJ63795-JPL CAAAJ63795-JPL
Yeast Hexokinase
Enzymes
  • Hygroscopic
Catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing ATP as a phosphate source. Used to determine glucose, fructose, mannose and ATP.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty