Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bovine Hemoglobin

Supplier: Thermo Scientific
J63838-06 J63838-14 J63838-22 J63838-22 J63838-14
AAJ63838-22EA 398.02 USD InStock
AAJ63838-22 CAAAJ63838-22 CAAAJ63838-14 AAJ63838-06 AAJ63838-14
Bovine Hemoglobin
Proteins and Peptides
Hemoglobin is the most important respiratory protein of vertebrates by virtue of its ability to transport oxygen from the lungs to body tissues, and to facilitate the return transport of carbon dioxide.

  • Dark Red to Brown Lyophilized Powder, 64.5 kDa
  • Store at Cold Temperatures
  • Soluble in water

Used as a coloring agent for pet foods, a natural source of iron for nutraceuticals, a protein source for non-ruminant animals, and as a raw material for pharmaceutical porphyrin derivative production.

Caution: Incompatible with oxidizng agents and heat.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty