Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trypsin inhibitor, chicken egg whites

Supplier: Thermo Scientific
J63927-03 J63927-04 J63927-ME J63927-ME J63927-04 J63927-03
AAJ63927-04EA 474.73 USD InStock
AAJ63927-04 CAAAJ63927-ME AAJ63927-03 AAJ63927-ME CAAAJ63927-03 CAAAJ63927-04
Trypsin inhibitor, chicken egg whites
Proteins and Peptides
Powder

CAS No.: 9035-81-8
Physical Form: Powder
Appearance: White
MDL No.: MFCD00082102

Applications: Inhibits the pancreatic enzyme trypsin

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty