Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Bromoguanosine

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Amino-8-bromo-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one

J63940-06 J63940-06
AAJ63940-06EA 136.96 USD InStock
AAJ63940-06 CAAAJ63940-06
8-Bromoguanosine
8-Bromoguanosine
Powder


Formula: C₁₀H₁₂BrN₅O₅
MW: 362.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037985
CAS Number: 4016-63-1
EINECS: 223-677-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty