Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Calcitonin Gene Related Peptide II

Supplier: Thermo Scientific
J64037-EXD J64037-EXD
AAJ64037-EXDEA 232.33 USD InStock
AAJ64037-EXD CAAAJ64037-EXD
Human Calcitonin Gene Related Peptide II
Proteins and Peptides
CAS No.: 98824-26-1
Molecular Formula: C162H267N51O48S3
Formula Weight: 3793.35
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00130617

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty