Synonyms:
Deacetylated chitin
, Poly(D-glucosamine)
J64143-18
J64143-30
J64143-36
J64143-30
J64143-36
J64143-18
AAJ64143-30EA
295.88
USD
InStock
CAAAJ64143-36
CAAAJ64143-18
AAJ64143-30
AAJ64143-18
AAJ64143-36
CAAAJ64143-30
Chitosan ≥85%, deacetylated
Chitosan
Chitosan is used as a flocculant. It also finds use in protein precipitation and as an encapsulating agent.
Powder