Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-β-D-Arabinofuranosyluracil 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Ara-UUracil 1-beta-D-Arabinofuranoside

J64147-06 J64147-06 J64147-03 J64147-14 J64147-03 J64147-14
AAJ64147-06EA 46.04 USD InStock
AAJ64147-06 AAJ64147-03 AAJ64147-14 CAAAJ64147-06 CAAAJ64147-03 CAAAJ64147-14
1-β-D-Arabinofuranosyluracil 99%
1-beta-D-Arabinofuranosyluracil
Powder


Formula: C₉H₁₂N₂O₆
MW: 244.20 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065998
CAS Number: 3083-77-0
EINECS: 221-386-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty