Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Azacytidine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Amino-1-(beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-oneLadakamycin

J64159-06 J64159-06 J64159-03 J64159-03
AAJ64159-06EA 291.09 USD InStock
AAJ64159-06 CAAAJ64159-06 AAJ64159-03 CAAAJ64159-03
5-Azacytidine 98%
5-Azacytidine
Causes DNA demethylation or hemi-demethylation; a powerful mutagen

Powder


Formula: C₈H₁₂N₄O₅
MW: 244.21 g/mol
Melting Pt: 226…232 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00006539
CAS Number: 320-67-2
EINECS: 206-280-2
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty