Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cytidine-5'-diphosphocholine sodium salt (CDP choline), hydrate

Supplier: Thermo Scientific Chemicals
Synonyms: Citicoline sodium saltCDP-choline sodium saltCDP-Choline

J64161-09 J64161-14 J64161-14 J64161-09 J64161-18 J64161-18
AAJ64161-18EA 253.63 USD InStock
AAJ64161-18 CAAAJ64161-09 AAJ64161-09 CAAAJ64161-18 CAAAJ64161-14 AAJ64161-14
Cytidine-5'-diphosphocholine sodium salt (CDP choline), hydrate
Cytidine-5'-diphosphocholine sodium salt (CDP choline)
Intermediate in the synthesis of phosphatidylcholine; also a phospholipase inhibitor

Powder


Formula: C₁₄H₂₆N₄NaO₁₁P₂·xH₂O
MW: 510.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150508
CAS Number: 33818-15-4
EINECS: 251-689-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty