Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-2'-deoxyadenosine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: (2R,3S,5R)-5-(2,6-Diamino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol

J64266-06 J64266-14 J64266-03 J64266-06 J64266-14 J64266-03
AAJ64266-14EA 145.05 USD InStock
AAJ64266-14 AAJ64266-03 CAAAJ64266-14 CAAAJ64266-03 CAAAJ64266-06 AAJ64266-06
2-Amino-2'-deoxyadenosine 99%
2-Amino-2'-deoxyadenosine
Powder


Formula: C₁₀H₁₄N₆O₃
MW: 266.261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047240
CAS Number: 4546-70-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty