Synonyms:
DNA sodium salt
, Thymonucleic acid sodium salt
J64400-03
J64400-MC
J64400-03
J64400-MC
AAJ64400-03EA
442.34
USD
InStock
CAAAJ64400-MC
CAAAJ64400-03
AAJ64400-03
AAJ64400-MC
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from calf thymus)
DNA sodium salt (Deoxyribonucleic acid sodium salt)
Deoxyribonucleic acid sodium salt, is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations.
- White Lyophilized powder
- Soluble in water
- Store at cold temperatures
Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. It was used as a standard to determine DNA content in gels and for DNA quantification by fluorescent assay.
Highly polymerized. Hyperchromicity is at least 27%.