Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-Deoxyadenosine-5'-monophosphate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: ((2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate

J64402-06 J64402-03 J64402-03 J64402-06
AAJ64402-03EA 101.66 USD InStock
CAAAJ64402-03 CAAAJ64402-06 AAJ64402-03 AAJ64402-06
2'-Deoxyadenosine-5'-monophosphate 98%
2'-Deoxyadenosine-5'-monophosphate
Crystalline powder


Formula: C₁₀H₁₄N₅O₆P
MW: 331.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005753
CAS Number: 653-63-4
EINECS: 211-503-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty