Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Cysteine methyl ester hydrochloride

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: H-Cys-OMe·HCl(R)-Methyl 2-amino-3-mercaptopropanoate hydrochloride

J64436-22 J64436-22 J64436-14 J64436-30 J64436-14 J64436-30
AAJ64436-14EA 67.96 USD InStock
CAAAJ64436-30 AAJ64436-14 AAJ64436-22 CAAAJ64436-22 AAJ64436-30 CAAAJ64436-14
L-Cysteine methyl ester hydrochloride
L-Cysteine methyl ester hydrochloride
Crystalline powder


Formula: C₄H₉NO₂S·HCl
MW: 171.7 g/mol
Melting Pt: 142 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00038998
CAS Number: 18598-63-5
EINECS: 242-435-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty