Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-6-chloropurine riboside 95%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Amino-6-chloro-9-(beta-D-ribofuranosyl)purine

J64468-06 J64468-06 J64468-03 J64468-03
AAJ64468-03EA 115.65 USD InStock
CAAAJ64468-06 CAAAJ64468-03 AAJ64468-03 AAJ64468-06
2-Amino-6-chloropurine riboside 95%
2-Amino-6-chloropurine riboside
Powder


Formula: C₁₀H₁₂ClN₅O₄
MW: 301.69 g/mol
Melting Pt: 165…167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005735
CAS Number: 2004-07-1
EINECS: 217-905-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty