Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Bromoadenosine-3',5'-cyclic monophosphate

Supplier: Thermo Scientific Chemicals

J64469-MF J64469-MC J64469-MD J64469-MC J64469-MF J64469-MD
AAJ64469-MFEA 188.67 USD InStock
CAAAJ64469-MD CAAAJ64469-MC AAJ64469-MF AAJ64469-MC AAJ64469-MD CAAAJ64469-MF
8-Bromoadenosine-3',5'-cyclic monophosphate
8-Bromoadenosine-3',5'-cyclic monophosphate
Powder


Formula: C₁₀H₁₁BrN₅O₆P
MW: 408.105 g/mol
Melting Pt: 254 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075580
CAS Number: 23583-48-4
EINECS: 245-760-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty