Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bolton-Hunter reagent (Sulfo-SHPP)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(4-Hydroxy-phenyl)-propionic acid-OSu

J64508-03 J64508-03
AAJ64508-03EA 190.79 USD InStock
AAJ64508-03 CAAAJ64508-03
Bolton-Hunter reagent (Sulfo-SHPP)
Bolton-Hunter reagent (Sulfo-SHPP)
For iodination of proteins that do not contain tyrosine residues

Powder

Applications: For iodination of proteins that do not contain tyrosine residues


Formula: C₁₃H₁₃NO₅
MW: 263.249 g/mol
Melting Pt: 132…133 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005515
CAS Number: 34071-95-9
EINECS: 251-818-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty