Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-Amino-2'-deoxyguanosine 98%

Supplier: Thermo Scientific Chemicals
Warning

J64650-MD J64650-03 J64650-03
10815-310EA 185.64 USD InStock
CAAAJ64650-03 10815-310 AAJ64650-03
2'-Amino-2'-deoxyguanosine 98%
2'-Amino-2'-deoxyguanosine
Powder


Formula: C₁₀H₁₄N₆O₄
Storage Temperature: Refrigerator
MDL Number: MFCD01723954
CAS Number: 60966-26-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty